Tetrathiafulvalene, 98.5%, Thermo Scientific Chemicals
Tetrathiafulvalene, 98.5%, Thermo Scientific Chemicals
Tetrathiafulvalene, 98.5%, Thermo Scientific Chemicals
Tetrathiafulvalene, 98.5%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Tetrathiafulvalene, 98.5%, Thermo Scientific Chemicals
CAS: 31366-25-3 | C6H4S4 | 204.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Packaging:
Glass bottle
Catalog number ACR179410010
Price (MYR)
1,327.00
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,327.00
Add to cart
Specifications
Chemical Name or MaterialTetrathiafulvalene
CAS31366-25-3
Melting Point117°C to 120°C
ColorBrown to Orange
EINECS Number250-593-7