Cellulose triacetate
Cellulose triacetate
Cellulose triacetate
Cellulose triacetate
Thermo Scientific Chemicals
Cellulose triacetate
CAS: 9012-09-3 | (C24H32O16)n
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
500 g
Packaging:
Plastic bottle
Catalog number ACR177822500
Price (MYR)
703.00
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
703.00
Add to cart
Chemical Identifiers
CAS541-53-7
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms
Appearance (Color)White to pale cream
Assay (HPLC)≥96.0%