Cellulose acetate, average M.W. 100,000
Cellulose acetate, average M.W. 100,000
Cellulose acetate, average M.W. 100,000
Cellulose acetate, average M.W. 100,000
Thermo Scientific Chemicals
Cellulose acetate, average M.W. 100,000
CAS: 9004-35-7 | (C6H7O5)nC2H3O
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
500 g
Packaging:
Plastic bottle
Catalog number ACR177780250
Price (MYR)
529.00
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
529.00
Add to cart
Chemical Identifiers
CAS67-63-0
IUPAC Namepropan-2-ol
Molecular FormulaC3H8O
InChI KeyKFZMGEQAYNKOFK-UHFFFAOYSA-N
SMILESCC(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Specific gravity(25°C) 0.783 to 0.787
Identification DPasses test
Non-volatile matter=<20 ppm
Water=<0.5 %
UVat 290 nm A: =<0.02