N-tert-Butyl-alpha-phenylnitrone, 98%, Thermo Scientific Chemicals
N-tert-Butyl-alpha-phenylnitrone, 98%, Thermo Scientific Chemicals
N-tert-Butyl-alpha-phenylnitrone, 98%, Thermo Scientific Chemicals
N-tert-Butyl-alpha-phenylnitrone, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N-tert-Butyl-alpha-phenylnitrone, 98%, Thermo Scientific Chemicals
CAS: 3376-24-7 | C11H15NO | 177.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number ACR177350050
Price (MYR)
2,149.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,149.00
Add to cart
Chemical Identifiers
CAS84228-93-3
IUPAC Name3-(pyridin-4-yl)prop-2-enoic acid
Molecular FormulaC8H7NO2
InChI KeySSAYTINUCCRGDR-UHFFFAOYSA-N
SMILESOC(=O)C=CC1=CC=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
Assay (Silylated GC)≥96.0%
Appearance (Color)White to cream to pale yellow