Carbon disulfide, 99.9%, extra pure, low in benzene content, Thermo Scientific Chemicals
Carbon disulfide, 99.9%, extra pure, low in benzene content, Thermo Scientific Chemicals
Carbon disulfide, 99.9%, extra pure, low in benzene content, Thermo Scientific Chemicals
Carbon disulfide, 99.9%, extra pure, low in benzene content, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Carbon disulfide, 99.9%, extra pure, low in benzene content, Thermo Scientific Chemicals
CAS: 75-15-0 | CS2 | 76.131 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
Packaging:
Glass Bottle
Catalog number ACR177280025
Price (MYR)
2,046.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
2,046.00
Add to cart
Chemical Identifiers
CAS6099-90-7
IUPAC Namebenzene-1,3,5-triol dihydrate
Molecular FormulaC6H10O5
InChI KeyMPYXTIHPALVENR-UHFFFAOYSA-N
SMILESO.O.OC1=CC(O)=CC(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (Silylated GC)≥97.5%
Identification (FTIR)Conforms
Water Content (Karl Fischer Titration)18-24%
Appearance (Color)White to cream to brown