Methanol, 99+%, extra pure, Thermo Scientific Chemicals
Methanol, 99+%, extra pure, Thermo Scientific Chemicals
Methanol, 99+%, extra pure, Thermo Scientific Chemicals
Methanol, 99+%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methanol, 99+%, extra pure, Thermo Scientific Chemicals
CAS: 67-56-1 | CH4O | 32.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 L
5 L
Packaging:
View all
Plastic Can
Catalog number ACR177150250
Price (MYR)
2,084.00
Add to cart
Quantity:
25 L
Packaging:
Plastic Can
Request bulk or custom format
Price (MYR)
2,084.00
Add to cart
Chemical Identifiers
CAS2050-47-7
IUPAC Name1-bromo-4-(4-bromophenoxy)benzene
Molecular FormulaC12H8Br2O
InChI KeyYAWIAFUBXXPJMQ-UHFFFAOYSA-N
SMILESBrC1=CC=C(OC2=CC=C(Br)C=C2)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
Assay (GC)≥98.5%
Infrared spectrumConforms
Assay (special)98.5-101.5% (Bromine combustion titration)
FormPowder or chunks