Methanol, 99+%, extra pure, Thermo Scientific Chemicals
Methanol, 99+%, extra pure, Thermo Scientific Chemicals
Methanol, 99+%, extra pure, Thermo Scientific Chemicals
Methanol, 99+%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methanol, 99+%, extra pure, Thermo Scientific Chemicals
CAS: 67-56-1 | CH4O | 32.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 L
25 L
Packaging:
View all
Plastic Drum
Catalog number ACR177150050
Price (MYR)
755.00
Add to cart
Quantity:
5 L
Packaging:
Plastic Drum
Request bulk or custom format
Price (MYR)
755.00
Add to cart
Specifications
Chemical Name or MaterialMethanol
Melting Point-98.0°C
CAS67-56-1
Health Hazard 1Danger
Health Hazard 2GHS H Statement:
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.
Highly flammable liquid and vapor.