tert-Butyl methyl ether, 99%, pure, Thermo Scientific Chemicals
tert-Butyl methyl ether, 99%, pure, Thermo Scientific Chemicals
tert-Butyl methyl ether, 99%, pure, Thermo Scientific Chemicals
tert-Butyl methyl ether, 99%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
tert-Butyl methyl ether, 99%, pure, Thermo Scientific Chemicals
CAS: 1634-04-4 | C5H12O | 88.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
2.5 L
Packaging:
Glass Bottle
Catalog number ACR177042500
Price (MYR)
381.00
Add to cart
Quantity:
250 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
381.00
Add to cart
Chemical Identifiers
CAS696-62-8
IUPAC Name1-iodo-4-methoxybenzene
Molecular FormulaC7H7IO
InChI KeySYSZENVIJHPFNL-UHFFFAOYSA-N
SMILESCOC1=CC=C(I)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to brown
Appearance (Form)Crystalline powder and/or chunks
AppearanceMay darken on storage
Infrared spectrumConforms
Melting point46°C to 51°C