tert-Butyl methyl ether, 99%, pure, Thermo Scientific Chemicals
tert-Butyl methyl ether, 99%, pure, Thermo Scientific Chemicals
tert-Butyl methyl ether, 99%, pure, Thermo Scientific Chemicals
tert-Butyl methyl ether, 99%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
tert-Butyl methyl ether, 99%, pure, Thermo Scientific Chemicals
CAS: 1634-04-4 | C5H12O | 88.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
250 mL
Packaging:
Glass Bottle
Catalog number ACR177040025
Price (MYR)
617.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
617.00
Add to cart
Chemical Identifiers
CAS7440-31-5
IUPAC Nametin
Molecular FormulaSn
InChI KeyATJFFYVFTNAWJD-UHFFFAOYSA-N
SMILES[Sn]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silvery
Assay from Supplier's CofA≥99.9985% (metals basis)
FormFoil, 2.0mm (0.08in) thick.