Methanol, 99.9%, for analysis, Thermo Scientific Chemicals
Methanol, 99.9%, for analysis, Thermo Scientific Chemicals
Methanol, 99.9%, for analysis, Thermo Scientific Chemicals
Methanol, 99.9%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methanol, 99.9%, for analysis, Thermo Scientific Chemicals
CAS: 67-56-1 | CH4O | 32.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Catalog number ACR176840025
Price (MYR)
565.00
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (MYR)
565.00
Add to cart
Chemical Identifiers
CAS1710-98-1
IUPAC Name4-tert-butylbenzoyl chloride
Molecular FormulaC11H13ClO
InChI KeyWNLMYNASWOULQY-UHFFFAOYSA-N
SMILESCC(C)(C)C1=CC=C(C=C1)C(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥97.5%
FormLiquid
Refractive Index1.5340-1.5380 @ 20?C
For analysis
RUO – Research Use Only