Methanol, 99.9%, for analysis, Thermo Scientific Chemicals
Methanol, 99.9%, for analysis, Thermo Scientific Chemicals
Methanol, 99.9%, for analysis, Thermo Scientific Chemicals
Methanol, 99.9%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methanol, 99.9%, for analysis, Thermo Scientific Chemicals
CAS: 67-56-1 | CH4O | 32.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Catalog number ACR176840010
Price (MYR)
436.00
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
436.00
Add to cart
Chemical Identifiers
CAS63-68-3
IUPAC Name(2S)-2-amino-4-(methylsulfanyl)butanoic acid
Molecular FormulaC5H11NO2S
InChI KeyFFEARJCKVFRZRR-BYPYZUCNSA-N
SMILESCSCC[C@H](N)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder or crystals
Loss on drying=<0.5 % (1 g, 105°C)
Specific optical rotation+22.4° to +24.7° (20°C, 589 nm) (c=2, 6 N HCl)
TLC=<0.5 % of any individual impurity
View more
For analysis
RUO – Research Use Only