Cyclohexane, 99.5%, for analysis, Thermo Scientific Chemicals
Cyclohexane, 99.5%, for analysis, Thermo Scientific Chemicals
Cyclohexane, 99.5%, for analysis, Thermo Scientific Chemicals
Cyclohexane, 99.5%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cyclohexane, 99.5%, for analysis, Thermo Scientific Chemicals
CAS: 110-82-7 | C6H12 | 84.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
500 mL
1 L
Packaging:
Glass Bottle
Catalog number ACR176810025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS10326-27-9
IUPAC Namebarium(2+) dihydrate dichloride
Molecular FormulaBaCl2H4O2
InChI KeyPWHCIQQGOQTFAE-UHFFFAOYSA-L
SMILESO.O.[Cl-].[Cl-].[Ba++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or crystalline powder
Assay from Supplier's CofA≥99.0% BaCl₂.2H₂O
Heavy Metals(ICP-OES): ≤5ppm
Impurity contentCa: ≤0.05%
View more
For analysis
RUO – Research Use Only