Acetone, 99.5+%, for analysis, Thermo Scientific Chemicals
Acetone, 99.5+%, for analysis, Thermo Scientific Chemicals
Acetone, 99.5+%, for analysis, Thermo Scientific Chemicals
Acetone, 99.5+%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Acetone, 99.5+%, for analysis, Thermo Scientific Chemicals
CAS: 67-64-1 | C3H6O | 58.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
Packaging:
Glass Bottle
Plastic Bottle
Catalog number ACR176800025
Price (MYR)
695.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
695.00
Add to cart
Chemical Identifiers
CAS5396-89-4
IUPAC Namebenzyl 3-oxobutanoate
Molecular FormulaC11H12O3
InChI KeyWOFAGNLBCJWEOE-UHFFFAOYSA-N
SMILESCC(=O)CC(=O)OCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)≥96.0%
Refractive Index1.5100-1.5150 @ 20?C
Appearance (Color)Clear colorless to yellow to orange
For analysis
RUO – Research Use Only