4-Acetylbenzoic acid, 98%
4-Acetylbenzoic acid, 98%
4-Acetylbenzoic acid, 98%
4-Acetylbenzoic acid, 98%
Thermo Scientific Chemicals
4-Acetylbenzoic acid, 98%
CAS: 586-89-0 | C9H8O3 | 164.16 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR17586010010 gGlass bottle
Catalog number ACR175860100
Price (MYR)
1,149.00
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,149.00
Add to cart
Chemical Identifiers
CAS18942-49-9
IUPAC Name(2R)-2-{[(tert-butoxy)carbonyl]amino}-3-phenylpropanoic acid
Molecular FormulaC14H19NO4
InChI KeyZYJPUMXJBDHSIF-LLVKDONJSA-N
SMILESCC(C)(C)OC(=O)N[C@H](CC1=CC=CC=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Optical Rotation-24.5? ? 1.5? (c=1, ethanol)
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
FormCrystals or powder or crystalline powder
Melting Point (clear melt)82.5-88.5?C
Appearance (Color)White