4-Isopropylaniline, 99%, Thermo Scientific Chemicals
4-Isopropylaniline, 99%, Thermo Scientific Chemicals
4-Isopropylaniline, 99%, Thermo Scientific Chemicals
4-Isopropylaniline, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Isopropylaniline, 99%, Thermo Scientific Chemicals
CAS: 99-88-7 | C9H13N | 135.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
Packaging:
Glass bottle
Catalog number ACR174530500
Price (MYR)
559.00
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
559.00
Add to cart
Chemical Identifiers
CAS77-86-1
IUPAC Name2-amino-2-(hydroxymethyl)propane-1,3-diol
Molecular FormulaC4H11NO3
InChI KeyLENZDBCJOHFCAS-UHFFFAOYSA-N
SMILESNC(CO)(CO)CO
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder or crystals or crystalline powder
Assay from Supplier's CofA≥99.9%
Protease activityNone detected
RNaseNone detected
UV/VisA280 (1M, water): ≤0.05