Pentadecafluorooctanoic acid, 96%, Thermo Scientific Chemicals
Pentadecafluorooctanoic acid, 96%, Thermo Scientific Chemicals
Pentadecafluorooctanoic acid, 96%, Thermo Scientific Chemicals
Pentadecafluorooctanoic acid, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Pentadecafluorooctanoic acid, 96%, Thermo Scientific Chemicals
CAS: 335-67-1 | C8HF15O2 | 414.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Packaging:
Glass bottle
Catalog number ACR173960250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS20816-12-0
IUPAC Nametetraoxoosmium
Molecular FormulaO4Os
InChI KeyVUVGYHUDAICLFK-UHFFFAOYSA-N
SMILESO=[Os](=O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow
Assay (special)3.0 ?0.1%