Pentadecafluorooctanoic acid, 96%, Thermo Scientific Chemicals
Pentadecafluorooctanoic acid, 96%, Thermo Scientific Chemicals
Pentadecafluorooctanoic acid, 96%, Thermo Scientific Chemicals
Pentadecafluorooctanoic acid, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Pentadecafluorooctanoic acid, 96%, Thermo Scientific Chemicals
CAS: 335-67-1 | C8HF15O2 | 414.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number ACR173960050
Price (MYR)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1694-92-4
IUPAC Name2-nitrobenzene-1-sulfonyl chloride
Molecular FormulaC6H4ClNO4S
InChI KeyWPHUUIODWRNJLO-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=CC=C1S(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to brown
FormCrystals or powder or crystalline powder or fused solid
Assay (Titration ex Chloride)≥96.0 to ≤104.0%
Identification (FTIR)Conforms
Melting Point (clear melt)63.0-72.0?C