5-Nitro-2-furaldehyde, 97%, Thermo Scientific Chemicals
5-Nitro-2-furaldehyde, 97%, Thermo Scientific Chemicals
5-Nitro-2-furaldehyde, 97%, Thermo Scientific Chemicals
5-Nitro-2-furaldehyde, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
5-Nitro-2-furaldehyde, 97%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
Packaging:
Glass bottle
Catalog number ACR173860100
Price (MYR)
209.11
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
209.11
Add to cart
Chemical Identifiers
CAS1948-33-0
IUPAC Name2-tert-butylbenzene-1,4-diol
Molecular FormulaC10H14O2
InChI KeyBGNXCDMCOKJUMV-UHFFFAOYSA-N
SMILESCC(C)(C)C1=CC(O)=CC=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
AppearanceMay contain black particles
Melting point125°C to 130°C
Appearance (Form)Crystalline powder
Infrared spectrumConforms
GC>=96.0 %