o-Anisic acid, 99%, Thermo Scientific Chemicals
o-Anisic acid, 99%, Thermo Scientific Chemicals
o-Anisic acid, 99%, Thermo Scientific Chemicals
o-Anisic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
o-Anisic acid, 99%, Thermo Scientific Chemicals
CAS: 579-75-9 | C8H8O3 | 152.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Plastic bottle
Catalog number ACR173751000
Price (MYR)
467.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
467.00
Add to cart
Chemical Identifiers
CAS6152-67-6
IUPAC Name4-(phenylamino)benzene-1-sulfonic acid sodium
Molecular FormulaC12H11NNaO3S
InChI KeyVMVSDVIYIMNZBS-UHFFFAOYSA-N
SMILES[Na].OS(=O)(=O)C1=CC=C(NC2=CC=CC=C2)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to beige to light grey
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Sensitivity(as indicator) passes test