Phenyl disulfide, 99%, Thermo Scientific Chemicals
Phenyl disulfide, 99%, Thermo Scientific Chemicals
Phenyl disulfide, 99%, Thermo Scientific Chemicals
Phenyl disulfide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phenyl disulfide, 99%, Thermo Scientific Chemicals
CAS: 882-33-7 | C12H10S2 | 218.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Packaging:
Plastic bottle
Catalog number ACR173712500
Price (MYR)
1,445.00
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,445.00
Add to cart
Chemical Identifiers
CAS1390-65-4
IUPAC Name3,5,6,8-tetrahydroxy-1-methyl-9,10-dioxo-7-[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]-9,10-dihydroanthracene-2-carboxylic acid
Molecular FormulaC22H20O13
InChI KeyDGQLVPJVXFOQEV-UHFFFAOYNA-N
SMILESCC1=C(C(O)=O)C(O)=CC2=C1C(=O)C1=C(O)C(C3OC(CO)C(O)C(O)C3O)=C(O)C(O)=C1C2=O
View more
SpecificationsSpecification SheetSpecification Sheet
BSC CertificationPasses
Infrared spectrumConforms
UV spectrumComplies
Appearance (Form)Powder
Appearance (Color)Dark red