Phenyl disulfide, 99%, Thermo Scientific Chemicals
Phenyl disulfide, 99%, Thermo Scientific Chemicals
Phenyl disulfide, 99%, Thermo Scientific Chemicals
Phenyl disulfide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phenyl disulfide, 99%, Thermo Scientific Chemicals
CAS: 882-33-7 | C12H10S2 | 218.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Packaging:
Plastic bottle
Catalog number ACR173710500
Price (MYR)
546.00
Add to cart
Quantity:
50 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
546.00
Add to cart
Chemical Identifiers
CAS123-86-4
IUPAC Namebutyl acetate
Molecular FormulaC6H12O2
InChI KeyDKPFZGUDAPQIHT-UHFFFAOYSA-N
SMILESCCCCOC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
GC=<0.2 % (n-Butyl alcohol)
Titrable acid=<0.0016 meq/g
Water=<0.1 % (K.F.)
Residue after evaporation=<0.001 %
Substance darkened by H2SO4Passes test