4,4'-Biphenol, 97%, Thermo Scientific Chemicals
4,4'-Biphenol, 97%, Thermo Scientific Chemicals
4,4'-Biphenol, 97%, Thermo Scientific Chemicals
4,4'-Biphenol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4,4'-Biphenol, 97%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
10 g
Packaging:
View all
Plastic bottle
Catalog number ACR173671000
Price (MYR)
1,126.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,126.00
Add to cart
Chemical Identifiers
CAS50-30-6
IUPAC Name2,6-dichlorobenzoic acid
Molecular FormulaC7H4Cl2O2
InChI KeyMRUDNSFOFOQZDA-UHFFFAOYSA-N
SMILESOC(=O)C1=C(Cl)C=CC=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Melting point141°C to 145°C
Appearance (Color)White to beige
Infrared spectrumConforms
HPLC>=98.0 %