4,4'-Biphenol, 97%, Thermo Scientific Chemicals
4,4'-Biphenol, 97%, Thermo Scientific Chemicals
4,4'-Biphenol, 97%, Thermo Scientific Chemicals
4,4'-Biphenol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4,4'-Biphenol, 97%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
100 g
Packaging:
View all
Glass bottle
Catalog number ACR173670100
Price (MYR)
402.00
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
402.00
Add to cart
Chemical Identifiers
CAS56-40-6
IUPAC Name2-aminoacetic acid
Molecular FormulaC2H5NO2
InChI KeyDHMQDGOQFOQNFH-UHFFFAOYSA-N
SMILESNCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
Assay (Non-aqueous acid-base Titration)≥98.5 to ≤101.5% (non-U.S. specification)
CommentSpecification differs for U.S. and non-U.S. material where indicated
FormCrystals or powder or crystalline powder
Assay from Suppliers CofA≥98.5% (U.S. specification)