Celite™, for analysis, high purity analytical grade, Thermo Scientific Chemicals
Celite™, for analysis, high purity analytical grade, Thermo Scientific Chemicals
Celite™, for analysis, high purity analytical grade, Thermo Scientific Chemicals
Celite™, for analysis, high purity analytical grade, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Celite™, for analysis, high purity analytical grade, Thermo Scientific Chemicals
CAS: 61790-53-2 | O2Si | 60.083 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
10 g
Packaging:
Glass Bottle
Catalog number ACR173460250
Price (MYR)
1,311.00
Add to cart
Quantity:
25 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
1,311.00
Add to cart
Chemical Identifiers
CAS5995-86-8
IUPAC Name3,4,5-trihydroxybenzoate
Molecular FormulaC7H5O5
InChI KeyLNTHITQWFMADLM-UHFFFAOYSA-M
SMILESOC1=CC(=CC(O)=C1O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Sulfated ash=<0.1 %
Titration with NaOH>=99 % (On dry substance)
Appearance (Form)Powder
Infrared spectrumConforms
Loss on drying=<10 % (120°C)
View more
Filter aid
RUO – Research Use Only