Celite™, for analysis, high purity analytical grade, Thermo Scientific Chemicals
Celite™, for analysis, high purity analytical grade, Thermo Scientific Chemicals
Celite™, for analysis, high purity analytical grade, Thermo Scientific Chemicals
Celite™, for analysis, high purity analytical grade, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Celite™, for analysis, high purity analytical grade, Thermo Scientific Chemicals
CAS: 61790-53-2 | O2Si | 60.083 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
Packaging:
Glass Bottle
Catalog number ACR173460100
Price (MYR)
792.00
Add to cart
Quantity:
10 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
792.00
Add to cart
Chemical Identifiers
CAS6141-58-8
IUPAC Namemethyl 2-methylfuran-3-carboxylate
Molecular FormulaC7H8O3
InChI KeyUVRRIABXNIGUJZ-UHFFFAOYSA-N
SMILESCOC(=O)C1=C(C)OC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to brown
Assay (GC)≥96.0%
Refractive Index1.4710-1.4760 @ 20?C
FormLiquid
Filter aid
RUO – Research Use Only