ε-Caprolactone monomer, 99%, Thermo Scientific Chemicals
ε-Caprolactone monomer, 99%, Thermo Scientific Chemicals
ε-Caprolactone monomer, 99%, Thermo Scientific Chemicals
ε-Caprolactone monomer, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
ε-Caprolactone monomer, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
1 kg
Packaging:
Glass bottle
Catalog number ACR173440025
Price (MYR)
2,405.00
Add to cart
Quantity:
2.5 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,405.00
Add to cart
Chemical Identifiers
CAS101772-29-6
IUPAC Name2-{[(tert-butoxy)carbonyl](methyl)amino}-3-hydroxypropanoic acid
Molecular FormulaC9H17NO5
InChI KeyNJQGIDVCNBZXLG-UHFFFAOYNA-N
SMILESCN(C(CO)C(O)=O)C(=O)OC(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormSolid
Assay (unspecified)>96.0% (NMR)