ε-Caprolactone monomer, 99%, Thermo Scientific Chemicals
ε-Caprolactone monomer, 99%, Thermo Scientific Chemicals
ε-Caprolactone monomer, 99%, Thermo Scientific Chemicals
ε-Caprolactone monomer, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
ε-Caprolactone monomer, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
2.5 kg
Packaging:
Glass bottle
Catalog number ACR173440010
Price (MYR)
976.00
Add to cart
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
976.00
Add to cart
Specifications
Chemical Name or Materialε-Caprolactone monomer
CAS502-44-3
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes serious eye irritation.
Health Hazard 3GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.