Abietic acid, 85%, Thermo Scientific Chemicals
Abietic acid, 85%, Thermo Scientific Chemicals
Abietic acid, 85%, Thermo Scientific Chemicals
Abietic acid, 85%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Abietic acid, 85%, Thermo Scientific Chemicals
CAS: 514-10-3 | C20H30O2 | 302.46 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number ACR173130250
Price (MYR)
682.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
682.00
Add to cart
Chemical Identifiers
CAS13419-61-9
IUPAC Namesodium decane-1-sulfonate
Molecular FormulaC10H21NaO3S
InChI KeyAIMUHNZKNFEZSN-UHFFFAOYSA-M
SMILES[Na+].CCCCCCCCCCS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Loss on drying=<5 % (100°C, 3 h) (vacuum)
Appearance (Color)White to cream
Appearance (Form)Crystalline powder
Titration after Ion exchange>=95 % (On dry substance)