3-O-Methyldopamine hydrochloride, 99+%, Thermo Scientific Chemicals
3-O-Methyldopamine hydrochloride, 99+%, Thermo Scientific Chemicals
3-O-Methyldopamine hydrochloride, 99+%, Thermo Scientific Chemicals
3-O-Methyldopamine hydrochloride, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-O-Methyldopamine hydrochloride, 99+%, Thermo Scientific Chemicals
CAS: 1477-68-5 | C9H13NO2·HCl | 203.67 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
1 g
Packaging:
Glass bottle
Catalog number ACR172851000
Price (MYR)
599.00
Add to cart
Quantity:
100 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
599.00
Add to cart
Specifications
Chemical Name or Material3-O-Methyldopamine hydrochloride
Name Notep.a.
CAS1477-68-5
CAS Min %99.0
Health Hazard 1GHS Signal Word: Warning