3-O-Methyldopamine hydrochloride, 99+%, Thermo Scientific Chemicals
3-O-Methyldopamine hydrochloride, 99+%, Thermo Scientific Chemicals
3-O-Methyldopamine hydrochloride, 99+%, Thermo Scientific Chemicals
3-O-Methyldopamine hydrochloride, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-O-Methyldopamine hydrochloride, 99+%, Thermo Scientific Chemicals
CAS: 1477-68-5 | C9H13NO2·HCl | 203.67 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
100 mg
Packaging:
Glass bottle
Catalog number ACR172850010
Price (MYR)
2,169.00
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,169.00
Add to cart
Chemical Identifiers
CAS78-78-4
IUPAC Name2-methylbutane
Molecular FormulaC5H12
InChI KeyQWTDNUCVQCZILF-UHFFFAOYSA-N
SMILESCCC(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Liquid
Refractive index1.3530 to 1.3550 (20°C, 589 nm)
Appearance (Color)Clear colorless
GC>=99.0 %