CAPS, 99%, for biochemistry
CAPS, 99%, for biochemistry
CAPS, 99%, for biochemistry
CAPS, 99%, for biochemistry
Thermo Scientific Chemicals
CAPS, 99%, for biochemistry
CAS: 1135-40-6 | C9H19NO3S | 221.32 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number ACR172621000
Price (MYR)
660.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
660.00
Add to cart
Chemical Identifiers
CAS1879-09-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Appearance (Form)Liquid or solid
Infrared spectrumConforms
GC>=96.0 %
Biological buffer
RUO – Research Use Only