4-Fluorobenzylamine, 97%, Thermo Scientific Chemicals
4-Fluorobenzylamine, 97%, Thermo Scientific Chemicals
4-Fluorobenzylamine, 97%, Thermo Scientific Chemicals
4-Fluorobenzylamine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Fluorobenzylamine, 97%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number ACR171950250
Price (MYR)
677.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
677.00
Add to cart
Chemical Identifiers
CAS7787-35-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark blue or dark green to black
FormPowder
Assay (unspecified)>88.0% ex Barium
Assay (unspecified)>88.0% ex Manganese