p-Toluenesulfonic acid monohydrate, 97.5%, pure, Thermo Scientific Chemicals
p-Toluenesulfonic acid monohydrate, 97.5%, pure, Thermo Scientific Chemicals
p-Toluenesulfonic acid monohydrate, 97.5%, pure, Thermo Scientific Chemicals
p-Toluenesulfonic acid monohydrate, 97.5%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
p-Toluenesulfonic acid monohydrate, 97.5%, pure, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number ACR171785000
Price (MYR)
393.00
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
393.00
Add to cart
Chemical Identifiers
CAS614-80-2
IUPAC NameN-(2-hydroxyphenyl)acetamide
Molecular FormulaC8H9NO2
InChI KeyADVGKWPZRIDURE-UHFFFAOYSA-N
SMILESCC(=O)NC1=CC=CC=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)Pale cream to cream to dark brown to brown
Assay (GC)≥96.0%
Melting Point (clear melt)204.0-214.0?C