p-Toluenesulfonic acid monohydrate, 97.5%, pure, Thermo Scientific Chemicals
p-Toluenesulfonic acid monohydrate, 97.5%, pure, Thermo Scientific Chemicals
p-Toluenesulfonic acid monohydrate, 97.5%, pure, Thermo Scientific Chemicals
p-Toluenesulfonic acid monohydrate, 97.5%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
p-Toluenesulfonic acid monohydrate, 97.5%, pure, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
500 g
5 kg
Packaging:
View all
Plastic bottle
Catalog number ACR171780010
Price (MYR)
503.00
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
503.00
Add to cart
Chemical Identifiers
CAS4431-00-9
IUPAC Nametriammonium 5-[(3-carboxylato-4-hydroxyphenyl)(3-carboxylato-4-oxocyclohexa-2,5-dien-1-yl)methyl]-2-hydroxybenzoate
Molecular FormulaC22H25N3O9
InChI KeyUDPQMNAHKKTHHI-UHFFFAOYNA-N
SMILES[NH4+].[NH4+].[NH4+].OC1=CC=C(C=C1C([O-])=O)C(C1C=CC(=O)C(=C1)C([O-])=O)C1=CC=C(O)C(=C1)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark red
Assay (Aqueous acid-base Titration)≥90%
FormPowder or crystalline powder
CommentSuitable for the estimation of aluminium