Cyclohexanepropionic acid, 99%, Thermo Scientific Chemicals
Cyclohexanepropionic acid, 99%, Thermo Scientific Chemicals
Cyclohexanepropionic acid, 99%, Thermo Scientific Chemicals
Cyclohexanepropionic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cyclohexanepropionic acid, 99%, Thermo Scientific Chemicals
CAS: 701-97-3 | C9H16O2 | 156.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
Catalog number ACR171450500
Price (MYR)
3,505.00
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (MYR)
3,505.00
Add to cart
Chemical Identifiers
CAS67-64-1
IUPAC Namepropan-2-one
Molecular FormulaC3H6O
InChI KeyCSCPPACGZOOCGX-UHFFFAOYSA-N
SMILESCC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=99.0 %
Appearance (Form)Clear liquid
Color scale=<10 APHA
Water=<0.5 % (K.F.)
Residue after evaporation=<0.005 %