3-Aminopyrazole, 98%, Thermo Scientific Chemicals
3-Aminopyrazole, 98%, Thermo Scientific Chemicals
3-Aminopyrazole, 98%, Thermo Scientific Chemicals
3-Aminopyrazole, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-Aminopyrazole, 98%, Thermo Scientific Chemicals
CAS: 1820-80-0 | C3H5N3 | 83.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
2.5 g
Packaging:
Glass bottle
Catalog number ACR171030100
Price (MYR)
1,499.00
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,499.00
Add to cart
Chemical Identifiers
CAS999-81-5
IUPAC Name(2-chloroethyl)trimethylazanium chloride
Molecular FormulaC5H13Cl2N
InChI KeyUHZZMRAGKVHANO-UHFFFAOYSA-M
SMILES[Cl-].C[N+](C)(C)CCCl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Infrared spectrumConforms
Water=<2.0 % (K.F.)
Appearance (Form)Crystalline powder or crystals and/or chunks
Titration with HClO497.5 to 102.5 %