3-Aminopyrazole, 98%, Thermo Scientific Chemicals
3-Aminopyrazole, 98%, Thermo Scientific Chemicals
3-Aminopyrazole, 98%, Thermo Scientific Chemicals
3-Aminopyrazole, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-Aminopyrazole, 98%, Thermo Scientific Chemicals
CAS: 1820-80-0 | C3H5N3 | 83.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 g
10 g
Packaging:
Glass bottle
Catalog number ACR171030025
Price (MYR)
663.00
Add to cart
Quantity:
2.5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
663.00
Add to cart
Specifications
Chemical Name or Material3-Aminopyrazole
CAS1820-80-0
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0