3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine-p,p'-disulfonic acid, disodium salt hydrat, Thermo Scientific Chemicals
3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine-p,p'-disulfonic acid, disodium salt hydrat, Thermo Scientific Chemicals
3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine-p,p'-disulfonic acid, disodium salt hydrat, Thermo Scientific Chemicals
3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine-p,p'-disulfonic acid, disodium salt hydrat, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine-p,p'-disulfonic acid, disodium salt hydrat, Thermo Scientific Chemicals
CAS: 1264198-47-1 | C20H12N4Na2O6S2 | 514.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number ACR171010010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS580-17-6
IUPAC Namequinolin-3-amine
Molecular FormulaC9H8N2
InChI KeySVNCRRZKBNSMIV-UHFFFAOYSA-N
SMILESNC1=CN=C2C=CC=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals
Melting point92°C to 96°C
Appearance (Color)White to beige to orange-brown
Infrared spectrumConforms
GC>=98.5 %