α-Bromo-p-toluic acid, 97%, Thermo Scientific Chemicals
α-Bromo-p-toluic acid, 97%, Thermo Scientific Chemicals
α-Bromo-p-toluic acid, 97%, Thermo Scientific Chemicals
α-Bromo-p-toluic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
α-Bromo-p-toluic acid, 97%, Thermo Scientific Chemicals
CAS: 6232-88-8 | C8H7BrO2 | 215.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Packaging:
Glass bottle
Catalog number ACR170400500
Price (MYR)
2,120.00
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,120.00
Add to cart
Specifications
Chemical Name or Materialα-Bromo-p-toluic acid
Name Note97%
CAS6232-88-8
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning