α-Bromo-p-toluic acid, 97%, Thermo Scientific Chemicals
α-Bromo-p-toluic acid, 97%, Thermo Scientific Chemicals
α-Bromo-p-toluic acid, 97%, Thermo Scientific Chemicals
α-Bromo-p-toluic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
α-Bromo-p-toluic acid, 97%, Thermo Scientific Chemicals
CAS: 6232-88-8 | C8H7BrO2 | 215.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number ACR170400100
Price (MYR)
704.00
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
704.00
Add to cart
Chemical Identifiers
CAS97-65-4
IUPAC Name2-methylidenebutanedioic acid
Molecular FormulaC5H6O4
InChI KeyLVHBHZANLOWSRM-UHFFFAOYSA-N
SMILESOC(=O)CC(=C)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Heavy metals (as Pb)=<20 ppm
Appearance (Form)Crystalline powder or crystals
Sulfate (SO4)=<300 ppm
Water=<0.5 %
Sulfated ash=<0.1 %