2-Bromoanisole, 97%, Thermo Scientific Chemicals
2-Bromoanisole, 97%, Thermo Scientific Chemicals
2-Bromoanisole, 97%, Thermo Scientific Chemicals
2-Bromoanisole, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Bromoanisole, 97%, Thermo Scientific Chemicals
CAS: 578-57-4 | C7H7BrO | 187.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
Glass bottle
Catalog number ACR170261000
Price (MYR)
2,179.00
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,179.00
Add to cart
Specifications
Chemical Name or Material2-Bromoanisole
Name Note97%
CAS578-57-4
CAS Min %96.0
CAS Max %100.0