n-Pentane, +99%, extra pure, Thermo Scientific Chemicals
n-Pentane, +99%, extra pure, Thermo Scientific Chemicals
n-Pentane, +99%, extra pure, Thermo Scientific Chemicals
n-Pentane, +99%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
n-Pentane, +99%, extra pure, Thermo Scientific Chemicals
CAS: 109-66-0 | C5H12 | 72.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass Bottle
Catalog number ACR170070025
Price (MYR)
809.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
809.00
Add to cart
Chemical Identifiers
CAS103-47-9
IUPAC Namesodium 2-(cyclohexylamino)ethane-1-sulfonate
Molecular FormulaC8H16NNaO3S
InChI KeyTXWKJOVDGFRVOG-UHFFFAOYSA-M
SMILES[Na+].[O-]S(=O)(=O)CCNC1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
Identification (FTIR)Conforms
Water Content (Karl Fischer Titration)≤0.5%
Appearance (Color)White