Phenoxyacetic acid, 98+%, Thermo Scientific Chemicals
Phenoxyacetic acid, 98+%, Thermo Scientific Chemicals
Phenoxyacetic acid, 98+%, Thermo Scientific Chemicals
Phenoxyacetic acid, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phenoxyacetic acid, 98+%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
Packaging:
Plastic bottle
Catalog number ACR169875000
Price (MYR)
634.00
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
634.00
Add to cart
Chemical Identifiers
CAS33512-26-4
IUPAC Namediethyl [(1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)methyl]phosphonate
Molecular FormulaC13H16NO5P
InChI KeyIUZMHUAHKBHJFY-UHFFFAOYSA-N
SMILESCCOP(=O)(CN1C(=O)C2=CC=CC=C2C1=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (GC)≥96.0%
Melting Point (clear melt)58.0-68.0?C