Diiodomethane, 99+%, stabilized with silver wire, Thermo Scientific Chemicals
Diiodomethane, 99+%, stabilized with silver wire, Thermo Scientific Chemicals
Diiodomethane, 99+%, stabilized with silver wire, Thermo Scientific Chemicals
Diiodomethane, 99+%, stabilized with silver wire, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Diiodomethane, 99+%, stabilized with silver wire, Thermo Scientific Chemicals
CAS: 75-11-6 | CH2I2 | 267.84 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Packaging:
Glass bottle
Catalog number ACR169831000
Price (MYR)
706.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
706.00
Add to cart
Chemical Identifiers
CAS16523-54-9
IUPAC Namechlorodicyclohexylphosphane
Molecular FormulaC12H22ClP
InChI KeyAKJFBIZAEPTXIL-UHFFFAOYSA-N
SMILESClP(C1CCCCC1)C1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)> 98.0%
Appearance (Color)Colorless