Diiodomethane, 99+%, stabilized with silver wire, Thermo Scientific Chemicals
Diiodomethane, 99+%, stabilized with silver wire, Thermo Scientific Chemicals
Diiodomethane, 99+%, stabilized with silver wire, Thermo Scientific Chemicals
Diiodomethane, 99+%, stabilized with silver wire, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Diiodomethane, 99+%, stabilized with silver wire, Thermo Scientific Chemicals
CAS: 75-11-6 | CH2I2 | 267.84 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number ACR169830250
Price (MYR)
468.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
468.00
Add to cart
Specifications
Chemical Name or MaterialDiiodomethane
CAS75-11-6
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Toxic if swallowed.
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.
CAS7440-22-4