4-(Benzyloxy)phenol, 98%, Thermo Scientific Chemicals
4-(Benzyloxy)phenol, 98%, Thermo Scientific Chemicals
4-(Benzyloxy)phenol, 98%, Thermo Scientific Chemicals
4-(Benzyloxy)phenol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-(Benzyloxy)phenol, 98%, Thermo Scientific Chemicals
CAS: 103-16-2 | C13H12O2 | 200.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
Packaging:
Glass bottle
Catalog number ACR169780500
Price (MYR)
574.00
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
574.00
Add to cart
Specifications
Chemical Name or Material4-(Benzyloxy)phenol
Name Note98%
CAS103-16-2
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning