Paraformaldehyde, 90%, pure, Thermo Scientific Chemicals
Paraformaldehyde, 90%, pure, Thermo Scientific Chemicals
Paraformaldehyde, 90%, pure, Thermo Scientific Chemicals
Paraformaldehyde, 90%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Paraformaldehyde, 90%, pure, Thermo Scientific Chemicals
CAS: 30525-89-4 | CH2O | 30.026 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
Packaging:
Plastic bottle
Catalog number ACR169650010
Price (MYR)
495.00
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
495.00
Add to cart
Chemical Identifiers
CAS598-78-7
IUPAC Name2-chloropropanoic acid
Molecular FormulaC3H5ClO2
InChI KeyGAWAYYRQGQZKCR-UHFFFAOYNA-N
SMILESCC(Cl)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless
Assay (GC)≥92.0%
Assay (Aqueous acid-base Titration)≥92.0 to ≤108.0%
Refractive Index1.4315-1.4385 @ 20?C