Methyl (triphenylphosphoranylidene)acetate, 98%, Thermo Scientific Chemicals
Methyl (triphenylphosphoranylidene)acetate, 98%, Thermo Scientific Chemicals
Methyl (triphenylphosphoranylidene)acetate, 98%, Thermo Scientific Chemicals
Methyl (triphenylphosphoranylidene)acetate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methyl (triphenylphosphoranylidene)acetate, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number ACR169560250
Price (MYR)
704.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
704.00
Add to cart
Chemical Identifiers
CAS134150-01-9
IUPAC Name(4-propylphenyl)boronic acid
Molecular FormulaC9H13BO2
InChI KeyWLCGYIWOKVWFLB-UHFFFAOYSA-N
SMILESCCCC1=CC=C(C=C1)B(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
for C-C Bond Formation (Olefination)
RUO – Research Use Only