Phosphorus tribromide, 99%, Thermo Scientific Chemicals
Phosphorus tribromide, 99%, Thermo Scientific Chemicals
Phosphorus tribromide, 99%, Thermo Scientific Chemicals
Phosphorus tribromide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phosphorus tribromide, 99%, Thermo Scientific Chemicals
CAS: 7789-60-8 | Br3P | 270.69 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
100 g
Packaging:
Glass bottle
Catalog number ACR169475000
Price (MYR)
1,406.00
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,406.00
Add to cart
Chemical Identifiers
CAS26570-48-9
IUPAC Nameethane-1,2-diol; prop-2-enoic acid
Molecular FormulaC5H10O4
InChI KeyFCDZZFLRLVQGEH-UHFFFAOYSA-N
SMILESOCCO.OC(=O)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow to pale cream
FormGranular solid
Assay from Supplier's CofA≥95.0%
CommentInhibitor: Report type and quantity
Molecular Weight8,000 (approx.)
C-X bond formation reagent
RUO – Research Use Only