Phosphorus tribromide, 99%, Thermo Scientific Chemicals
Phosphorus tribromide, 99%, Thermo Scientific Chemicals
Phosphorus tribromide, 99%, Thermo Scientific Chemicals
Phosphorus tribromide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phosphorus tribromide, 99%, Thermo Scientific Chemicals
CAS: 7789-60-8 | Br3P | 270.69 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
500 g
Packaging:
Glass bottle
Catalog number ACR169471000
Price (MYR)
668.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
668.00
Add to cart
Specifications
Boiling Point175.0°C
Chemical Name or MaterialPhosphorus tribromide
Melting Point-40.0°C
CAS7789-60-8
ColorBrown or Colorless to Yellow
View more
C-X bond formation reagent
RUO – Research Use Only